Ethyl-5-fluoro-1H-indole-2-carboxylate 98%
Lieferant: Thermo Scientific
Synonyme:
Ethyl 5-fluoro-1H-indole-2-carboxylate, Ethyl 5-Fluoroindole-2-carboxylate, 5-Fluoroindole-2-carboxylic acid ethyl ester
L14010.03
L14010.06
L14010.03EA
33.6
EUR
L14010.03
L14010.06
Ethyl-5-fluoro-1H-indole-2-carboxylate 98%
Ethyl-5-fluoro-1H-indole-2-carboxylate
Purity: 98%
CAS No.: 348-36-7
Molecular Formula: C11H10FNO2
Formula Weight: 207.21
Melting Point: 147-152°
Beilstein Reference: 170254
MDL No.: MFCD00152076
Formel:
C₁₁H₁₀FNO₂ Molecular Weight: 207,21 g/mol Schmelzpunkt: 147…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00152076 CAS-Nummer: 348-36-7 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...