(S)-(-)-2-Methylbutylamine ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
L14063.03
L14063.MD
L14063.03EA
177
EUR
L14063.03
L14063.MD
(S)-(-)-2-Methylbutylamine ≥98%
(S)-(-)-2-Methylbutylamine
Purity: 98+%
CAS No.: 34985-37-0
Molecular Formula: C5H13N
Formula Weight: 87.16
Flash Point: 3°(37°F)
Refractive Index: 1.4105
Boiling Point: 96-97°
Beilstein Reference: 1718807
Optical Rotation: -5.9° (neat)
UN No.: UN2733
MDL No.: MFCD00064430
Formel:
C₅H₁₃N Molecular Weight: 87,16 g/mol Siedepunkt: 96…97 °C Dichte: 0,756 Flammpunkt: 3 °C (37 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064430 CAS-Nummer: 34985-37-0 EINECS: 252-307-6 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...