7,8-Dihydroxy-4-methylcoumarin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Methyldaphnetin, 7,8-Dihydroxy-4-methyl-2H-chromen-2-one

L14069.ME L14069.03
L14069.MEEA 32.1 EUR
L14069.ME L14069.03
7,8-Dihydroxy-4-methylcoumarin 97%
7,8-Dihydroxy-4-methylcoumarin

Purity: 97% CAS No.: 2107-77-9 Molecular Formula: C10H8O4 Formula Weight: 192.17 Storage Temperature: 2°C to 8°C Melting Point: 241-244° Beilstein Reference: 177613 MDL No.: MFCD00016972

Formel: C₁₀H₈O₄
Molecular Weight: 192,17 g/mol
Schmelzpunkt: 241…244 °C
MDL: MFCD00016972
CAS-Nummer: 2107-77-9
EINECS: 218-290-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR