(-)-Menthyl acetate 98%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
L-Menthyl acetate, L-Menthol acetate, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl acetate, (R)-(-)-Menthyl acetate, (-)-Acetoxy-p-menthane, (5R)-2-Isopropyl-5-methylcyclohexyl acetate, (1R)-(−)-Menthyl acetate
L14095.03
L14095.MD
L14095.03EA
60.9
EUR
L14095.03
L14095.MD
(-)-Menthyl acetate 98%
(-)-Menthyl acetate
Purity: 98%
CAS No.: 2623-23-6
Molecular Formula: C12H22O2
Formula Weight: 198.31
Flash Point: 77°(170°F)
Refractive Index: 1.4470
Boiling Point: 229-231°
Beilstein Reference: 2208505
Optical Rotation: -80.5° (c=8 in benzene)
MDL No.: MFCD00001482
Formel:
C₁₂H₂₂O₂ Molecular Weight: 198.31 g/mol Siedepunkt: 229…231 °C Dichte: 0.925 Flammpunkt: 77 °C (170 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001482 CAS-Nummer: 2623-23-6 EINECS: 220-076-0 Merck Index: 14,5839 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...