Genkwanin, 7-methoxyapigenin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Hydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-chromen-4-one, 4',5-Dihydroxy-7-methoxyflavone

L14169.MB L14169.MC
L14169.MBEA 61.2 EUR
L14169.MB L14169.MC
Genkwanin, 7-methoxyapigenin 97%
Genkwanin, 7-methoxyapigenin

Purity: 97% CAS No.: 437-64-9 Molecular Formula: C16H12O5 Formula Weight: 284.27 Melting Point: 288-290° Beilstein Reference: 292549 MDL No.: MFCD00017452

Formel: C₁₆H₁₂O₅
Molecular Weight: 284,27 g/mol
Schmelzpunkt: 288…290 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017452
CAS-Nummer: 437-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR