Fisetin 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,7,3',4'-Tetrahydroxyflavone

L14207.03 L14207.MD
L14207.03EA 175 EUR
L14207.03 L14207.MD
Fisetin 96%
Fisetin

Purity: 97% CAS No.: 528-48-3 Molecular Formula: C15H10O6 Formula Weight: 286.24 Melting Point: 330° dec. Beilstein Reference: 292829 MDL No.: MFCD00006829

Formel: C₁₅H₁₀O₆
Molecular Weight: 286.24 g/mol
Schmelzpunkt: 330 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006829
CAS-Nummer: 528-48-3
EINECS: 208-434-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR