(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98%
Lieferant: Thermo Scientific
Gefahr
Synonyme: (-)-MTPA-Cl, (-)-Mosher's Acid Chloride, (R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
L14325.MF L14325.MD L14325.03
L14325.MFEA 46.2 EUR
L14325.MF L14325.MD L14325.03
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98%
(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Purity: 98+% CAS No.: 39637-99-5 Molecular Formula: C10H8ClF3O2 Formula Weight: 252.62 Storage Temperature: -30°C to -10°C Flash Point: 89°(192°F) Refractive Index: 1.4690 Boiling Point: 213-214° Beilstein Reference: 3591565 Optical Rotation: -137° (neat) UN No.: UN3265 MDL No.: MFCD00044400
Formel: C₁₀H₈ClF₃O₂
Molecular Weight: 252,62 g/mol
Siedepunkt: 213…214 °C
Dichte: 1,353
Flammpunkt: 89 °C (192 °F)
Lagertemperatur: Gefrierschrank
MDL: MFCD00044400
CAS-Nummer: 39637-99-5
UN: 3265
ADR: 8,II
Merck Index: 14,6280