(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98%
Lieferant: Thermo Scientific
Gefahr
Synonyme: (+)-Mosher's Acid Chloride, (+)-MTPA-Cl, (S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, (S)-3,3,3-Trifluoro-2-methoxy-2-phenylpropanoyl chloride
L14332.MF L14332.MD L14332.03
L14332.MFEA 41.2 EUR
L14332.MF L14332.MD L14332.03
(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98%
(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Purity: 98+% CAS No.: 20445-33-4 Molecular Formula: C10H8ClF3O2 Formula Weight: 252.62 Storage Temperature: -30°C to -10°C Flash Point: 89°(192°F) Refractive Index: 1.4690 Boiling Point: 213-214° Beilstein Reference: 3591564 Optical Rotation: +137° (c=6.4 in carbon tetrachloride) UN No.: UN3265 MDL No.: MFCD00067105
Formel: C₁₀H₈ClF₃O₂
Molecular Weight: 252,62 g/mol
Siedepunkt: 213…214 °C
Dichte: 1,353
Flammpunkt: 89 °C (192 °F)
Lagertemperatur: Gefrierschrank
MDL: MFCD00067105
CAS-Nummer: 20445-33-4
UN: 3265
ADR: 8,II
Merck Index: 14,6280