3',5-Dihydroxy-4',6,7-trimethoxyflavone 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6,7-dimethoxy-4H-chromen-4-one, Eupatorin
L14495.MB
L14495.MC
L14495.MBEA
241
EUR
L14495.MB
L14495.MC
3',5-Dihydroxy-4',6,7-trimethoxyflavone 97%
3',5-Dihydroxy-4',6,7-trimethoxyflavone
Purity: 97%
CAS No.: 855-96-9
Molecular Formula: C18H16O7
Formula Weight: 344.32
Melting Point: 190-192°
Beilstein Reference: 1355109
MDL No.: MFCD00016929
Formel:
C₁₈H₁₆O₇ Molecular Weight: 344,32 g/mol Schmelzpunkt: 190…192 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016929 CAS-Nummer: 855-96-9 Merck Index: 14,3901 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...