Luteolin tetramethylether 97%

Lieferant: Thermo Scientific

Synonyme: 5,7,3',4'-Tetramethoxyflavone, 2-(3,4-Dimethoxyphenyl)-5,7-dimethoxy-4H-chromen-4-one

L14496.MC L14496.ME
L14496.MCEA 70.1 EUR
L14496.MC L14496.ME
Luteolin tetramethylether 97%
Luteolin tetramethylether

Purity: 97% CAS No.: 855-97-0 Molecular Formula: C19H18O6 Formula Weight: 342.35 Melting Point: 193-197° Beilstein Reference: 336320 MDL No.: MFCD00017558

Formel: C₁₉H₁₈O₆
Molecular Weight: 342,35 g/mol
Schmelzpunkt: 193…197 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017558
CAS-Nummer: 855-97-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR