4-(2-Hydroxyhexafluoroisopropyl)benzoic acid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(1,1,1,3,3,3-Hexafluoro-2-hydroxyprop-2-yl)benzoic acid

L14554.03 L14554.06
L14554.03EA 71.8 EUR
L14554.03 L14554.06
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid 97%
4-(2-Hydroxyhexafluoroisopropyl)benzoic acid

Purity: 97% CAS No.: 16261-80-6 Molecular Formula: C10H6F6O3 Formula Weight: 288.15 Odour: Odourless Melting Point: 122-126° Beilstein Reference: 2148278 MDL No.: MFCD00236043

Formel: C₁₀H₆F₆O₃
Molecular Weight: 288,15 g/mol
Schmelzpunkt: 122…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236043
CAS-Nummer: 16261-80-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR