Benzyl-(S)-(-)-lactate 97%
Lieferant: Thermo Scientific
Synonyme:
Benzyl L-lactate, (S)-(-)-Lactic Acid Benzyl Ester, Benzyl (S)-Lactate
L14645.06
L14645.14
L14645.06EA
58.3
EUR
L14645.06
L14645.14
Benzyl-(S)-(-)-lactate 97%
Benzyl-(S)-(-)-lactate
Purity: 97%
CAS No.: 56777-24-3
Molecular Formula: C10H12O3
Formula Weight: 180.20
Flash Point:
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,20 g/mol Siedepunkt: 105…109 °C (0,1 mmHg) Dichte: 1,120 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00209546 CAS-Nummer: 56777-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...