1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine 97%

Lieferant: Thermo Scientific
Achtung

L14835.03 L14835.06
L14835.03EA 57.1 EUR
L14835.03 L14835.06
1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine 97%
1,4-Dichloro-5,6,7,8-tetrahydro-5,8-ethanophthalazine

Purity: 97% CAS No.: 202823-67-4 Molecular Formula: C10H10Cl2N2 Formula Weight: 229.11 Melting Point: 208-211° MDL No.: MFCD01075667

Formel: C₁₀H₁₀Cl₂N₂
Molecular Weight: 229,11 g/mol
Schmelzpunkt: 208…211 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075667
CAS-Nummer: 202823-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR