Ethyl-4-hydroxy-3-methoxycinnamat 98%
Lieferant: Thermo Scientific
Synonyme: 4-Hydroxy-3-methoxycinnamic Acid Ethyl Ester, Ethyl 3-(4-hydroxy-3-methoxyphenyl)acrylate, Ethyl Ferulate, Ferulic Acid Ethyl Ester
L14960.03 L14960.MD
L14960.03EA 111 EUR
L14960.03 L14960.MD
Ethyl-4-hydroxy-3-methoxycinnamat 98%
Ethyl-4-hydroxy-3-methoxycinnamate
Purity: 99% CAS No.: 4046-02-0 Molecular Formula: C12H14O4 Formula Weight: 222.24 Melting Point: 56-58° Beilstein Reference: 2696807 MDL No.: MFCD00009190
Formel: C₁₂H₁₄O₄
Molecular Weight: 222,24 g/mol
Schmelzpunkt: 56…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009190
CAS-Nummer: 4046-02-0
EINECS: 223-745-5