Boran-Morpholin-Komplex 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Morpholine Borane
L15110.06
L15110.14
L15110.06EA
23.8
EUR
L15110.06
L15110.14
Boran-Morpholin-Komplex 97%
Borane-morpholine complex
Purity: 97%
CAS No.: 4856-95-5
Molecular Formula: C4H12BNO
Formula Weight: 100.96
Odour: Amine-like
Melting Point: 93-95°
UN No.: UN2925
MDL No.: MFCD00066736
Formel:
C₄H₁₂BNO Molecular Weight: 100,96 g/mol Schmelzpunkt: 93…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066736 CAS-Nummer: 4856-95-5 EINECS: 225-450-7 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...