(tert-Butoxycarbonylmethylene)triphenylphosphorane 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: tert-Butyl 2-(triphenylphosphoranylidene)acetate

L15144.06 L15144.14
L15144.06EA 74.9 EUR
L15144.06 L15144.14
(tert-Butoxycarbonylmethylene)triphenylphosphorane 98%
(tert-Butoxycarbonylmethylene)triphenylphosphorane

Purity: 98% CAS No.: 35000-38-5 Molecular Formula: C24H25O2P Formula Weight: 376.44 Melting Point: 146-150° Beilstein Reference: 2759875 UN No.: UN2811 MDL No.: MFCD00075545

Formel: C₂₄H₂₅O₂P
Molecular Weight: 376,436 g/mol
Schmelzpunkt: 146…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075545
CAS-Nummer: 35000-38-5
EINECS: 412-880-7
UN: 3464
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR