Diethylentriamin-pentaessigsäure-dianhydrid 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
DTPA Dianhydride, DTPA anhydride, N,N-Bis[2-(2,6-dioxomorpholino)ethyl]glycine
L15178.03
L15178.06
L15178.03EA
52.7
EUR
L15178.03
L15178.06
Diethylentriamin-pentaessigsäure-dianhydrid 95%
Diethylenetriaminepentaacetic dianhydride
Purity: 95%
CAS No.: 23911-26-4
Molecular Formula: C14H19N3O8
Formula Weight: 357.32
Melting Point: ca 190° dec.
Beilstein Reference: 1230954
MDL No.: MFCD00010697
Formel:
C₁₄H₁₉N₃O₈ Molecular Weight: 357,32 g/mol Schmelzpunkt: ∼ 190 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010697 CAS-Nummer: 23911-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...