N-Benzylglycinhydrochlorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Bzl-Gly-OH·HCl, Bzl-Gly-OH.HCl
L15200.14
L15200.22
L15200.14EA
118
EUR
L15200.14
L15200.22
N-Benzylglycinhydrochlorid ≥98%
N-Benzylglycine hydrochloride
Purity: 98+%
CAS No.: 7689-50-1
Molecular Formula: C6H5CH2NHCH2CO2H.HCl
Formula Weight: 201.65
Melting Point: ca 220° dec.
Beilstein Reference: 3915652
MDL No.: MFCD00156934
Formel:
C₉H₁₁NO₂·HCl Molecular Weight: 201.7 g/mol Schmelzpunkt: ∼ 220 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156934 CAS-Nummer: 7689-50-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...