3-Aminophthalhydrazide monosodium salt ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Sodium 5-amino-1,4-dioxo-3,4-dihydro-1H-phthalazin-2-ide, Luminol sodium salt
L15205.03
L15205.06
L15205.03EA
61
EUR
L15205.03
L15205.06
3-Aminophthalhydrazide monosodium salt ≥98%
3-Aminophthalhydrazide monosodium salt
Purity: 98+%
CAS No.: 20666-12-0
Molecular Formula: C8H6N3NaO2
Formula Weight: 199.15
MDL No.: MFCD00161491
Formel:
C₈H₆N₃NaO₂ Molecular Weight: 199.15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00161491 CAS-Nummer: 20666-12-0 EINECS: 629-344-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...