Methyl isopropyl carbonate 97%

Lieferant: Thermo Fisher Scientific
Achtung

L15314.14 L15314.03 L15314.06
L15314.14EA 307 EUR
L15314.14 L15314.03 L15314.06
Methyl isopropyl carbonate 97%
Methyl isopropyl carbonate

Purity: 97% CAS No.: 51729-83-0 Molecular Formula: C5H10O3 Formula Weight: 118.13 Flash Point: 27°(80°F) Refractive Index: 1.3830 Boiling Point: 116-118° Beilstein Reference: 1744735 UN No.: UN3272 MDL No.: MFCD01075689

Formel: C₅H₁₀O₃
Siedepunkt: 116…118 °C
Dichte: 0,975
Flammpunkt: 27 °C (80 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD01075689
CAS-Nummer: 51729-83-0
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR