Pyrrole-2-carbonitrile 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L15330.03
L15330.06
L15330.03EA
46.6
EUR
L15330.03
L15330.06
Pyrrole-2-carbonitrile 99%
Pyrrole-2-carbonitrile
Purity: 99%
CAS No.: 4513-94-4
Molecular Formula: C5H4N2
Formula Weight: 92.10
Refractive Index: 1.5500
Boiling Point: 92-94°/2mm
Beilstein Reference: 107047
UN No.: UN3276
MDL No.: MFCD00234061
Formel:
C₅H₄N₂ Molecular Weight: 92.101 g/mol Siedepunkt: 92…94 °C (2 mmHg) Dichte: 1.086 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234061 CAS-Nummer: 4513-94-4 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...