Bzl-Gly-OH ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-(Benzylamino)acetic acid, N-Benzylglycine

L15368.14 L15368.06
L15368.14EA 218 EUR
L15368.14 L15368.06
Bzl-Gly-OH ≥98%
Bzl-Gly-OH

Purity: 98+% CAS No.: 17136-36-6 Molecular Formula: C6H5CH2NHCH2CO2H Formula Weight: 165.19 Melting Point: ca 210° dec. Beilstein Reference: 2209788 MDL No.: MFCD00190772

Formel: C₉H₁₁NO₂
Molecular Weight: 165.19 g/mol
Schmelzpunkt: ∼ 210 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00190772
CAS-Nummer: 17136-36-6
EINECS: 241-194-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR