L-Pipecolsäure ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(S)-(-)-2-Piperidinecarboxylic Acid, H-HomoPro-OH, (S)-Piperidine-2-carboxylic acid, L-Pipecolinic Acid, L-Homoproline
L15373.03
L15373.06
L15373.03EA
115
EUR
L15373.03
L15373.06
L-Pipecolsäure ≥99%
L-Pipecolsäure
Purity: 99%
CAS No.: 3105-95-1
Molecular Formula: C6H11NO2
Formula Weight: 129.16
Melting Point: ca 270° dec.
Beilstein Reference: 81093
Optical Rotation: -26° (c=1 in water)
MDL No.: MFCD00005981
Formel:
C₆H₁₁NO₂ Molecular Weight: 129,16 g/mol Schmelzpunkt: ∼ 270 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005981 CAS-Nummer: 3105-95-1 EINECS: 221-462-1 Merck Index: 14,7458 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...