2,3-Dihydro-1H-benz[de]isoquinoline 97%
Lieferant: Thermo Scientific
|
Gefahr
|
L15613.03
L15613.MD
L15613.03EA
162
EUR
L15613.03
L15613.MD
2,3-Dihydro-1H-benz[de]isoquinoline 97%
2,3-Dihydro-1H-benz[de]isoquinoline
Purity: 97%
CAS No.: 22817-26-1
Molecular Formula: C12H11N
Formula Weight: 169.23
Melting Point: 96-101°
Boiling Point: 120°/0.5mm
Beilstein Reference: 134303
UN No.: UN3259
MDL No.: MFCD01075706
Formel:
C₁₂H₁₁N Molecular Weight: 169,23 g/mol Siedepunkt: 120 °C (0,5 mmHg) Schmelzpunkt: 96…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075706 CAS-Nummer: 22817-26-1 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...