L-Prolinbenzylester -hydrochlorid 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
H-Pro-OBzl.HCl
L15618.03
L15618.06
L15618.03EA
26.6
EUR
L15618.03
L15618.06
L-Prolinbenzylester -hydrochlorid 98%
L-Prolinbenzylester -hydrochlorid
Purity: 98%
CAS No.: 16652-71-4
Molecular Formula: C12H15NO2.HCl
Formula Weight: 241.72
Melting Point: 146-148°
Beilstein Reference: 3598081
Optical Rotation: -44° (c=1 in methanol)
MDL No.: MFCD00039002
Formel:
C₁₂H₁₅NO₂ Molecular Weight: 241.72 g/mol Schmelzpunkt: 146…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039002 CAS-Nummer: 16652-71-4 EINECS: 240-700-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...