[Hydroxy(tosyloxy)iodo]benzene 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Koser Reagent, Phenyliodosohydroxy Tosylate
L15701.06
L15701.14
L15701.06EA
46.1
EUR
L15701.06
L15701.14
[Hydroxy(tosyloxy)iodo]benzene 97%
[Hydroxy(tosyloxy)iodo]benzene
Purity: 97%
CAS No.: 27126-76-7
Molecular Formula: C13H13IO4S
Formula Weight: 392.22
Melting Point: 131-136°
Beilstein Reference: 2150074
MDL No.: MFCD00011547
Formel:
C₁₃H₁₃IO₄S Molecular Weight: 392,21 g/mol Schmelzpunkt: 131…136 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011547 CAS-Nummer: 27126-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...