4-Amino-2-isopropyl-5-methylphenol hydrochloride 97%
Lieferant: Thermo Scientific
|
Achtung
|
L16282.03
L16282.06
L16282.03EA
21.8
EUR
L16282.03
L16282.06
4-Amino-2-isopropyl-5-methylphenol hydrochloride 97%
4-Amino-2-isopropyl-5-methylphenol hydrochloride
Purity: 97%
CAS No.: 6321-11-5
Molecular Formula: C10H15NO.HCl
Formula Weight: 201.70
Odour: Odourless
Melting Point: 258-262°
Beilstein Reference: 3700651
MDL No.: MFCD00035504
Formel:
C₁₀H₁₆ClNO Schmelzpunkt: 258…262 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00035504 CAS-Nummer: 6321-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...