Ethyl-3-methylisoxazole-4-carboxylate 97%

Lieferant: Thermo Fisher Scientific

L16362.03 L16362.06
L16362.03EA 36.6 EUR
L16362.03 L16362.06
Ethyl-3-methylisoxazole-4-carboxylate 97%
Ethyl-3-methylisoxazole-4-carboxylate

Purity: 97% CAS No.: 20328-15-8 Molecular Formula: C7H9NO3 Formula Weight: 155.15 Refractive Index: 1.4595 Boiling Point: 64°/0.6mm Beilstein Reference: 1072756 MDL No.: MFCD01075736

Formel: C₇H₉NO₃
Molecular Weight: 155.15 g/mol
Siedepunkt: 64 °C (0.6 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD01075736
CAS-Nummer: 20328-15-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR