α-Methyl-4-trifluoromethylbenzyl alcohol 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-[4-(Trifluoromethyl)phenyl]ethanol

L16878.03 L16878.06
L16878.03EA 32.7 EUR
L16878.03 L16878.06
α-Methyl-4-trifluoromethylbenzyl alcohol 96%
alpha-Methyl-4-trifluoromethylbenzyl alcohol

Purity: 97% CAS No.: 1737-26-4 Molecular Formula: C9H9F3O Formula Weight: 190.17 Flash Point: 98°(208°F) Refractive Index: 1.4580 Boiling Point: 62-63°/0.5mm Beilstein Reference: 2257808 MDL No.: MFCD00039618

Formel: C₉H₉F₃O
Molecular Weight: 190,17 g/mol
Siedepunkt: 62…63 °C (0,5 mmHg)
Dichte: 1,237
Flammpunkt: 98 °C (208 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00039618
CAS-Nummer: 1737-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR