3-Bromo-2-chloropyridine-5-sulphonyl chloride 96%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 5-Bromo-6-chloropyridine-3-sulphonyl chloride

L17015.03 L17015.06
L17015.03EA 47.3 EUR
L17015.03 L17015.06
3-Bromo-2-chloropyridine-5-sulphonyl chloride 96%
3-Bromo-2-chloropyridine-5-sulphonyl chloride

Purity: 96% CAS No.: 216394-05-7 Molecular Formula: C5H2BrCl2NO2S Formula Weight: 290.95 Melting Point: 65-70° UN No.: UN3261 MDL No.: MFCD01318107

Formel: C₅H₂BrCl₂NO₂S
Molecular Weight: 290,96 g/mol
Schmelzpunkt: 65…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318107
CAS-Nummer: 216394-05-7
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR