4-Chlorbutylether 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4,4'-Dichlorodibutyl Ether, 4,4'-Dichlorobutyl Ether, 4-Chlorobutyl Ether

L17055.22 L17055.06 L17055.14
L17055.22EA 303 EUR
L17055.22 L17055.06 L17055.14
4-Chlorbutylether 99%
Bis(4-chlorobutyl)ether

Purity: 99% CAS No.: 6334-96-9 Molecular Formula: C8H16Cl2O Formula Weight: 199.12 Refractive Index: 1.4580 Boiling Point: 129-131°/10mm Beilstein Reference: 1739861 MDL No.: MFCD00019006

Formel: C₈H₁₆Cl₂O
Molecular Weight: 199,12 g/mol
Siedepunkt: 129…131 °C (10 mmHg)
Dichte: 1,081
Lagertemperatur: Raumtemperatur
MDL: MFCD00019006
CAS-Nummer: 6334-96-9
EINECS: 228-713-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR