1-Phenyl-1,2,3-butanetrione-2-oxime ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L17073.03
L17073.06
L17073.03EA
40.8
EUR
L17073.03
L17073.06
1-Phenyl-1,2,3-butanetrione-2-oxime ≥98%
1-Phenyl-1,2,3-butanetrione-2-oxime
Purity: 98+%
CAS No.: 6797-44-0
Molecular Formula: C10H9NO3
Formula Weight: 191.19
Odour: Odourless
Melting Point: 128-130°
Beilstein Reference: 1946580
MDL No.: MFCD00466576
Formel:
C₁₀H₉NO₃ Schmelzpunkt: 128…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00466576 CAS-Nummer: 6797-44-0 EINECS: 229-869-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...