Hydrogen fluoride pyridine complex (∼70% HF)
Lieferant: Thermo Scientific
Gefahr
Synonyme: Olah′s reagent, Pyridinium polybifluoride, Pyridine hydrofluoride, HF-Pyridine, PPHF, Poly(pyridine fluoride), Pyridinium poly(hydrogen fluoride)
L17117.14 L17117.22
L17117.14EA 42.6 EUR
L17117.14 L17117.22
Hydrogen fluoride pyridine complex (∼70% HF)
Hydrogen fluoride pyridine complex
CAS No.: 62778-11-4 Storage Temperature: 2°C to 8°C Boiling Point: 50°/1mm Beilstein Reference: 4750151 UN No.: UN1790 MDL No.: MFCD00012436
Formel: C₅H₅N·(HF)x
Siedepunkt: 50 °C (1 mmHg)
Dichte: 1,100
Lagertemperatur: Kühlschrank
MDL: MFCD00012436
CAS-Nummer: 62778-11-4
UN: 1790
ADR: 8,I
Merck Index: 14,6821