5-Methyl-1H-indole-2-carboxylic acid 99%

Lieferant: Thermo Fisher Scientific
Achtung

L17145.03 L17145.06
L17145.03EA 71.1 EUR
L17145.03 L17145.06
5-Methyl-1H-indole-2-carboxylic acid 99%
5-Methyl-1H-indole-2-carboxylic acid

Purity: 99% CAS No.: 10241-97-1 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Melting Point: ca 235° dec. Beilstein Reference: 144055 MDL No.: MFCD00047166

Formel: C₁₀H₉NO₂
Molecular Weight: 175.19 g/mol
Schmelzpunkt: ∼ 235 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00047166
CAS-Nummer: 10241-97-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR