N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine 90%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Chloro-N,N-diethyl-1,1,2-trifluoroethylamine
L17330.06
L17330.14
L17330.22
L17330.06EA
39.5
EUR
L17330.06
L17330.14
L17330.22
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine 90%
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine
Purity: 90%
CAS No.: 357-83-5
Molecular Formula: C6H11ClF3N
Formula Weight: 189.61
Storage Temperature: 2°C to 8°C
Boiling Point: 38°/10mm
Beilstein Reference: 1098675
UN No.: UN3267
MDL No.: MFCD00054672
Formel:
C₆H₁₁ClF₃N Molecular Weight: 189.61 g/mol Siedepunkt: 38 °C (10 mmHg) Dichte: 1.190 |
MDL:
MFCD00054672 CAS-Nummer: 357-83-5 ADR: 8 II |