5-Isopropyl-2-methoxybenzeneboronic acid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (5-Isopropyl-2-methoxyphenyl)boronic acid

L17460.03 L17460.06
L17460.03EA 47.4 EUR
L17460.03 L17460.06
5-Isopropyl-2-methoxybenzeneboronic acid ≥98%
5-Isopropyl-2-methoxybenzeneboronic acid

Purity: 98+% CAS No.: 216393-63-4 Molecular Formula: C10H15BO3 Formula Weight: 194.04 Melting Point: 85-87° Beilstein Reference: 9193266 MDL No.: MFCD01318154

Formel: C₁₀H₁₅BO₃
Molecular Weight: 194,04 g/mol
Schmelzpunkt: 85…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318154
CAS-Nummer: 216393-63-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR