4-Nitrophenyl-trifluormethansulfonat 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Trifluoromethanesulphonic acid 4-Nitrophenyl Ester, 4-Nitrophenyl Triflate

L17481.03 L17481.06
L17481.03EA 24.1 EUR
L17481.03 L17481.06
4-Nitrophenyl-trifluormethansulfonat 99%
4-Nitrophenyl-trifluormethansulfonat

Purity: 99% CAS No.: 17763-80-3 Molecular Formula: C7H4F3NO5S Formula Weight: 271.17 Storage Temperature: 2°C to 8°C Melting Point: 50-54° Beilstein Reference: 1998336 UN No.: UN2923 MDL No.: MFCD00192354

Formel: C₇H₄F₃NO₅S
Molecular Weight: 271,17 g/mol
Schmelzpunkt: 50…54 °C
MDL: MFCD00192354
CAS-Nummer: 17763-80-3
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR