Ethyl-4-acetyl-5-oxohexanoat 98%
Lieferant: Thermo Scientific
Synonyme:
Ethyl 4,4-Diacetylbutyrate, Ethyl 4-acetyl-5-oxohexanoate, 4,4-Diacetylbutyric Acid Ethyl Ester, 4-Acetyl-5-oxohexanoic Acid Ethyl Ester
L17496.06
L17496.14
L17496.06EA
14.2
EUR
L17496.06
L17496.14
Ethyl-4-acetyl-5-oxohexanoat 98%
Ethyl-4-acetyl-5-oxohexanoate
Purity: 98%
CAS No.: 2832-10-2
Molecular Formula: C10H16O4
Formula Weight: 200.24
Flash Point:
Formel:
C₁₀H₁₆O₄ Molecular Weight: 200,23 g/mol Siedepunkt: 153…154 °C (19 mmHg) Dichte: 1,067 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009211 CAS-Nummer: 2832-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...