5-Fluoro-1H-indole-3-carbaldehyde 98%

Lieferant: Thermo Fisher Scientific
Achtung

L17507.03 L17507.06 L17507.MD
L17507.03EA 126 EUR
L17507.03 L17507.06 L17507.MD
5-Fluoro-1H-indole-3-carbaldehyde 98%
5-Fluoro-1H-indole-3-carbaldehyde

Purity: 98% CAS No.: 2338-71-8 Molecular Formula: C9H6FNO Formula Weight: 163.15 Melting Point: 164° Beilstein Reference: 1449216 MDL No.: MFCD00022719

Formel: C₉H₆FNO
Schmelzpunkt: 164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022719
CAS-Nummer: 2338-71-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR