Fomblin® YR-1800

Lieferant: Thermo Scientific

Synonyme: Perfluoropolyether YR-1800

L17548.06 L17548.14
L17548.06EA 34.9 EUR
L17548.06 L17548.14
Fomblin® YR-1800
1-Propene, 1,1,2,3,3,3-hexafluoro-, oxidized, polymerized

CAS No.: 69991-67-9 Refractive Index: 1.3040 MDL No.: MFCD01323324

Dichte: 1,920
Lagertemperatur: Raumtemperatur
MDL: MFCD01323324
CAS-Nummer: 69991-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR