Ethyl-5-methoxyindole-2-carboxylate 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 5-Methoxyindole-2-carboxylic acid ethyl ester

L17596.03 L17596.06
L17596.03EA 35.3 EUR
L17596.03 L17596.06
Ethyl-5-methoxyindole-2-carboxylate 98%
Ethyl-5-methoxyindole-2-carboxylate

Purity: 98% CAS No.: 4792-58-9 Molecular Formula: C12H13NO3 Formula Weight: 219.24 Melting Point: 154-157° Beilstein Reference: 178104 MDL No.: MFCD00047163

Formel: C₁₂H₁₃NO₃
Schmelzpunkt: 154…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047163
CAS-Nummer: 4792-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR