5-(tert-Butoxycarbonylamino)-1-pentanol 96%
Lieferant: Thermo Scientific
Synonyme:
N-(5-Hydroxypentyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(5-Hydroxypentyl)carbamate, tert-Butyl (5-hydroxypentyl)carbamate, N-Boc-5-aminopentanol, 5-(Boc-amino)-1-pentanol
L17649.03
L17649.06
L17649.03EA
54.2
EUR
L17649.03
L17649.06
5-(tert-Butoxycarbonylamino)-1-pentanol 96%
5-(tert-Butoxycarbonylamino)-1-pentanol
Purity: 96%
CAS No.: 75178-90-4
Molecular Formula: C10H21NO3
Formula Weight: 203.28
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₁₀H₂₁NO₃ Molecular Weight: 203,28 g/mol Dichte: 1,000 Flammpunkt: >109 °C (228 °F) |
MDL:
MFCD01862955 CAS-Nummer: 75178-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...