3,5-Bis(trifluoromethyl)phenyldimethylsilane 95%

Lieferant: Thermo Scientific
Achtung

L17673.03 L17673.06
L17673.03EA 54.7 EUR
L17673.03 L17673.06
3,5-Bis(trifluoromethyl)phenyldimethylsilane 95%
3,5-Bis(trifluoromethyl)phenyldimethylsilane

Purity: 95% CAS No.: 33558-36-0 Molecular Formula: C10H10F6Si Formula Weight: 272.27 Refractive Index: 1.4125 Boiling Point: 40°/1.5mm Beilstein Reference: 2859742 MDL No.: MFCD02093948

Formel: C₁₀H₁₀F₆Si
Molecular Weight: 272,27 g/mol
Siedepunkt: 40 °C (1,5 mmHg)
Dichte: 1,230
Lagertemperatur: Raumtemperatur
MDL: MFCD02093948
CAS-Nummer: 33558-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR