Ethyl-5-propyl-1H-pyrazole-3-carboxylate 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Ethyl 5-Propylpyrazole-3-carboxylate
L17687.14
L17687.06
L17687.14EA
359
EUR
L17687.14
L17687.06
Ethyl-5-propyl-1H-pyrazole-3-carboxylate 98%
Ethyl-5-propyl-1H-pyrazole-3-carboxylate
Purity: 98%
CAS No.: 92945-27-2
Molecular Formula: C9H14N2O2
Formula Weight: 182.22
Melting Point: 47-51°
Beilstein Reference: 7634535
MDL No.: MFCD02093950
Formel:
C₉H₁₄N₂O₂ Molecular Weight: 182.22 g/mol Schmelzpunkt: 47…51 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08700607 CAS-Nummer: 92945-27-2 |