Ethyl-4-acetylbutyrate 97%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
Ethyl 4-Acetylbutyrate
L17848.22
L17848.06
L17848.14
L17848.22EA
310
EUR
L17848.22
L17848.06
L17848.14
Ethyl-4-acetylbutyrate 97%
Ethyl-4-acetylbutyrate
Purity: 97%
CAS No.: 13984-57-1
Molecular Formula: C8H14O3
Formula Weight: 158.20
Flash Point: 69°(156°F)
Refractive Index: 1.4275
Boiling Point: 65°/0.76mm
Beilstein Reference: 1762625
MDL No.: MFCD00009213
Formel:
C₈H₁₄O₃ Molecular Weight: 158.20 g/mol Siedepunkt: 65 °C (0.76 mmHg) Dichte: 0.989 Flammpunkt: 69 °C (156 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009213 CAS-Nummer: 13984-57-1 EINECS: 237-776-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...