Benzyl-5-amino-1H-pyrazole-4-carboxylate ≥98%

Lieferant: Thermo Scientific
Achtung

L17859.03 L17859.06
L17859.03EA 15.6 EUR
L17859.03 L17859.06
Benzyl-5-amino-1H-pyrazole-4-carboxylate ≥98%
Benzyl-5-amino-1H-pyrazole-4-carboxylate

Purity: 98+% CAS No.: 32016-28-7 Molecular Formula: C11H11N3O2 Formula Weight: 217.23 Melting Point: 121-123° Beilstein Reference: 791498 MDL No.: MFCD03426023

Formel: C₁₁H₁₁N₃O₂
Molecular Weight: 217,23 g/mol
Schmelzpunkt: 121…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426023
CAS-Nummer: 32016-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR