3-(Bromomethyl)phenoxyacetic acid 97%

Lieferant: Thermo Scientific
Gefahr

L18029.06 L18029.03
L18029.06EA 143 EUR
L18029.06 L18029.03
3-(Bromomethyl)phenoxyacetic acid 97%
3-(Bromomethyl)phenoxyacetic acid

Purity: 97% CAS No.: 136645-25-5 Molecular Formula: C9H9BrO3 Formula Weight: 245.07 Melting Point: 124-128° UN No.: UN3261 MDL No.: MFCD02093985

Formel: C₉H₉BrO₃
Molecular Weight: 245,07 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093985
CAS-Nummer: 136645-25-5
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR