3-(4-Methoxybenzoyl)acrylic acid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(4-Methoxyphenyl)-4-oxobut-2-enoic acid

L18071.06 L18071.03
L18071.06EA 175 EUR
L18071.06 L18071.03
3-(4-Methoxybenzoyl)acrylic acid 98%
3-(4-Methoxybenzoyl)acrylic acid

Purity: 98% CAS No.: 5711-41-1 Molecular Formula: C11H10O4 Formula Weight: 206.20 Melting Point: 137-140° Beilstein Reference: 474529 MDL No.: MFCD00020486

Formel: C₁₁H₁₀O₄
Molecular Weight: 206,20 g/mol
Schmelzpunkt: 137…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00020486
CAS-Nummer: 5711-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR