(5R)-5-(Hydroxymethyl)-2-pyrrolidinon 99%

Lieferant: Thermo Scientific
Achtung

L18358.03 L18358.06
L18358.03EA 49.9 EUR
L18358.03 L18358.06
(5R)-5-(Hydroxymethyl)-2-pyrrolidinon 99%
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone

Purity: 99% CAS No.: 66673-40-3 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Melting Point: 83-85° Boiling Point: 147-149°/0.06m Beilstein Reference: 5728422 Optical Rotation: -30° (c=2 in ethanol) MDL No.: MFCD00077791

Formel: C₅H₉NO₂
Molecular Weight: 115,13 g/mol
Siedepunkt: 147…149 °C (0,06m)
Schmelzpunkt: 83…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077791
CAS-Nummer: 66673-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR