1-Acetylisonipecotoyl chloride 97% may contain up to ca 1M free HCl

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-Acetylpiperidine-4-carbonyl chloride

L18426.03 L18426.06
L18426.03EA 77.1 EUR
L18426.03 L18426.06
1-Acetylisonipecotoyl chloride 97% may contain up to ca 1M free HCl
1-Acetylisonipecotoyl chloride

Purity: 97% CAS No.: 59084-16-1 Molecular Formula: C8H12ClNO2 Formula Weight: 189.64 Melting Point: 131-133° Beilstein Reference: 389571 UN No.: UN3261 MDL No.: MFCD02094017

Formel: C₈H₁₂ClNO₂
Molecular Weight: 189,64 g/mol
Schmelzpunkt: 131…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02094017
CAS-Nummer: 59084-16-1
EINECS: 261-594-7
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR